Learn More
Macherey-Nagel™ Polygram CEL 400-10 Glass Plates
2285.00 NOK - 3170.00 NOK
Specifications
Type Glass Plate
Thickness (Metric) 0.1 mm
View More Specs
Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
10226911
View Documents
Macherey-Nagel™
808072
10 x 20 cm
3170.00 NOK
Pack of 50
Only null left
In Stock
Add to basket
10236191
View Documents
Macherey-Nagel™
808073
20 x 20 cm
2285.00 NOK
Pack of 25
Only null left
In Stock
Specifications
Glass Plate
0.1 mm
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Polygram CEL 400-10 Glass Plates