missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 60 to 20 C8

Brand:  Macherey-Nagel™ 711008.100

 View more versions of this product

Product Code. 10125171

  • 2435.00 NOK / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

10 to 15 μm
POLYGOPREP 60 to 20
2 to 8
POLYGOPREP C8
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
100 g
350 m2/g
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Polygoprep™ 60 to 20 C8 > Quantity: 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.