Learn More
Macherey-Nagel™ Polygoprep™ 100 to 12
Brand:  Macherey-Nagel™ 711002.1000
 View more versions of this product
Product Code. 10620332
  • 4145.00 NOK / 1000g
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
10 to 15 μm
POLYGOPREP 100 to 12
2 to 8
POLYGOPREP
Polygoprep
Liquid Chromatography
100 Å
Liquid Chromatography
1 kg
280 m2/g
Silica
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Product Certifications
Ofertas especiales
Ofertas especiales