Learn More
Masterflex™ Polyethylene Straight Connector
Designed for quick and easy assembly without tools
280.00 NOK - 348.00 NOK
Specifications
Description Barb Connection
Autoclavable Not Autoclavable
Material HDPE
Sterilization Method Ethylene Oxide
Color Translucent
View More Specs
Products 3
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
11765559
View Documents
Masterflex™
30621-79
5/8 in.
280.00 NOK
Pack of 10
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
11775559
View Documents
Masterflex™
30621-81
3/4 in.
324.00 NOK
Pack of 10
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
11770049
View Documents
Masterflex™
30621-83
1 in.
348.00 NOK
Pack of 10
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
Description
Description
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
Barb Connection
HDPE
Translucent
Not Autoclavable
Ethylene Oxide
Straight Connector
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Masterflex™ Polyethylene Straight Connector