Learn More
Kartell™ Plastilab™ Polyethylene Slide Mailer
Designed with support bars for strength and safety in transporting specimen slides.
Brand:  Kartell™ 0092500
Product Code. 12024857
  • 233.00 NOK / Pack of 10
Only null left
Estimated Shipment: 21-07-2025
Description
Designed with support bars for strength and safety in transporting specimen slides.
  • Slides with own compartments with support bar at 55mm centers
  • For protection 76 x 26mm specimen slides
  • Useful for cytological and geological specimens
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Plastilab™ Polyethylene Slide Mailer > Width: 84mm