Learn More
Ansell Edmont™ Polyethylene Shoecovers
For low-cost protection in light duty applications. Ansell Edmont™ Polyethylene Shoecovers are a good choice for single-use tasks.
Brand: Ansell Edmont™ SC41BCPE
562.64 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
This item is not returnable.
View return policy
Description
- Cool, light and comfortable to wear
- Manufactured from blue chlorinated polyethylene
- With elastic opening
Specifications
Polyethylene | |
100/Pack | |
Shoe Covers |
Blue | |
15 x 41cm |
Product Content Correction
Product Title
Ansell Edmont™ Polyethylene Shoecovers > Dimensions (L x W): 15 x 41cm; Quantity: 100/Pk.