Learn More
Resch Glas™ Polyethylene Rolled Edge Injection Vials
Click to view available options
Capacity (Metric):
50 mL
Specifications
Specifications
Capacity (Metric) | 50 mL |
Color | Clear |
Includes | Snap-On Lid |
Material | Polyethylene |
No. per Pack | 1 Each |
Quantity | 1 Each |
Weight (Metric) | 5.62 kg |
Clarity | Clear |
Closure Included | Yes |
Shape | Round |
Show More |
Product Content Correction
Product Title
Resch Glas™ Polyethylene Rolled Edge Injection Vials >