Learn More
Kartell™ Polyethylene Disc Membrane
These discs are made of polyethylene and have good chemical resistance. Kartell™ Polyethylene Disc Membrane is used to separate the filter paper from the funnel plate when working with a Buchner Funnel.
Brand:  Kartell™ 0083900
116.85 NOK valid until 2025-12-31
Use promo code "24321" to get your promotional price.
 View more versions of this product
Product Code. 10011051
  • 123.00 NOK / Pack of 10
Only null left
Estimated Shipment: 03-07-2025
Description
  • Useful interface between filter paper and perforated Buchner plate
  • Avoids clogging of filter holes
Specifications
Specifications
Screen Filter
Polyethylene
Interface between filter paper and perforated Buchner plate
Circle
Disc
300 ÎĽm
10/Pack
90 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Polyethylene Disc Membrane > Diameter: 90 mm