Learn More
Brady™ Polyester Labels for the BBP33/i3300 Printer
Our B-494 polyester label is a reliable sample identification and traceability solution.
1260.00 NOK - 2925.00 NOK
Specifications
Adhesive Type | Acrylic |
---|---|
Finish | Gloss |
Format | Roll |
Material | Polyester |
Printable Sides | Front |
Product Code | Brand | Length (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Color | Price | Quantity & Availability | ||||
17170521
|
Brady™
307007 |
9.53 mm | Yellow, White |
1260.00 NOK
Pack of 1500 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
17180521
|
Brady™
307008 |
9.53 mm | Red, White |
1260.00 NOK
Pack of 1500 |
In Stock
Log in to see stock. |
||||
17190521
|
Brady™
307009 |
9.53 mm | Pink, White |
1260.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17100531
|
Brady™
307010 |
9.53 mm | Orange, White |
1260.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17110531
|
Brady™
307011 |
9.53 mm | Green, White |
1260.00 NOK
Pack of 1500 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
17120531
|
Brady™
307012 |
9.53 mm | Blue, White |
1260.00 NOK
Pack of 1500 |
In Stock
Log in to see stock. |
||||
17110521
|
Brady™
307001 |
12.70 mm | Yellow, White |
2925.00 NOK
Pack of 2500 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
17120521
|
Brady™
307002 |
12.70 mm | Red, White |
2925.00 NOK
Pack of 2500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17130521
|
Brady™
307003 |
12.70 mm | Pink, White |
2925.00 NOK
Pack of 2500 |
In Stock
Log in to see stock. |
||||
17140521
|
Brady™
307004 |
12.70 mm | White |
2925.00 NOK
Pack of 2500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17150521
|
Brady™
307005 |
12.70 mm | Green, White |
2925.00 NOK
Pack of 2500 |
In Stock
Log in to see stock. |
||||
17160521
|
Brady™
307006 |
12.70 mm | Blue, White |
2925.00 NOK
Pack of 2500 |
In Stock
Log in to see stock. |
||||
17130531
|
Brady™
307019 |
15.24 mm | Yellow, White |
2465.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17140531
|
Brady™
307020 |
15.24 mm | Red, White |
2465.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17150531
|
Brady™
307021 |
15.24 mm | Pink, White |
2465.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17160531
|
Brady™
307022 |
15.24 mm | Orange, White |
2465.00 NOK
Pack of 1500 |
In Stock
Log in to see stock. |
||||
17170531
|
Brady™
307023 |
15.24 mm | Green, White |
2465.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17180531
|
Brady™
307024 |
15.24 mm | Blue, White |
2465.00 NOK
Pack of 1500 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
Description
- The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
- Thanks to subsurface printing, the label's color code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Acrylic | |
Roll | |
Front | |
Labels | |
BBP™33 Label Printer; BradyPrinter i3300 Industrial Label Printer | |
-320.8°F to +158°F | |
0.0762 mm |
Gloss | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
-196°C | |
-196°C to +70°C |