missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Polycarbonate Protective Safety Glasses, 2750 Series
Click to view available options
Lens Tint:
Clear
Gray
Description
Provide additional features and style for increased security, comfort, and wearer acceptance.
- Excellent impact resistance
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Adjustable temple length to suit the individual
- Anti-scratch and anti-fog coating for increased durability and improved vision
- Ideal for use in many environments including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking

Specifications
Specifications
Certifications/Compliance | EN 166:2001 |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety glasses; Safety Spectacles |
For Use With (Application) | Construction,‎ forestry |
Frame Color | Black |
No. per Pack | 1 Ea. |
Temple Style | Flat, Adjustable length |
Lens Material | Polycarbonate |
Documents
Scientific Resources
Flyers
3M 2750 Series Safety Spectacles Flyer
File size: 228.8 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Polycarbonate Protective Safety Glasses, 2750 Series >
Spot an opportunity for improvement?Share a Content Correction