Learn More
Cole-Parmer™ Polyamide Tweezer
Use these general purpose tweezers for a more precise grip. Cole-Parmer™ Polyamide Tweezers are serrated with rounded tips.
Brand:  Cole-Parmer™ 06446-20
Product Code. 11769669
  • 200.00 NOK / Each
Only null left
Estimated Shipment: 11-07-2025
Description
Non-sterile yellow tweezers for general lab use.
Specifications
Specifications
Round
Polyamide
Tweezers
Tweezer
Each
180 mm
Each
Round
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Polyamide Tweezer > Length: 180mm