Learn More
VITLAB™ Polyamide Spatulas
Manufactured from Polyamide. VITLAB™ Polyamide Spatulas can be used for a wide range of laboratory purposes.
Brand: VITLAB™ 80593
This item is not returnable.
View return policy
Description
- Glass-fiber reinforced
- Spatula spoon shape
- Easy to hold handle
Specifications
Spoon | |
Red | |
Polyamide | |
210 mm |
Product Content Correction
Product Title
VITLAB™ Polyamide Spatulas > Length: 210mm