Learn More
Macherey-Nagel™ Polyamide CC 6
4860.00 NOK - 24585.00 NOK
Product Code | Brand | Quantity | Particle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Particle Size | Price | Quantity & Availability | |||||
10033091
|
Macherey-Nagel™
815610.1 |
1 kg | <0.07 |
5400.00 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10328411
|
Macherey-Nagel™
815600.1 |
1 kg | 0.10 to 0.3 mm |
4860.00 NOK
1kg |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
10441862
|
Macherey-Nagel™
815620.5 |
5 kg | 0.05 to 0.16 mm |
24585.00 NOK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10286231
|
Macherey-Nagel™
815600.5 |
5 kg | 0.10 to 0.3 mm |
22125.00 NOK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Polyamide CC 6 | |
Polyamide |
Product Title
Macherey-Nagel™ Polyamide CC 6