Learn More
MP Biomedicals™ Poly-L-Glutamic Acid, MW 50,000-100,000
Brand:  MP Biomedicals™ 0215191891
 View more versions of this product
Product Code. 11447162
  • 14360.00 NOK / 1g
Explore more special offers
Specifications
95%
Powder
50000 to 100000
1000 mg
0°C
Used in the construction of Polypeptide based Copolymer membranes, micelles and nanoparticles
Poly-L-Glutamic Acid from Synthetic