Learn More
POLR2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20370-1-AP-20UL
 View more versions of this product
Product Code. 16859904
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The antibody is specific to POLR2B.
This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.
Specifications
Specifications
POLR2B
Polyclonal
Unconjugated
POLR2B
hRPB140, hsRPB2, POL2RB, POLR2B, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RPB2
Rabbit
Antigen Affinity Chromatography
RUO
231329, 289561, 5431
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30876, Q8CFI7
Polr2b
Peptide
20 μL
Primary
Human, Mouse, Rat