missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ PMMA (Plexiglas) Two-Tier Test Tube Racks

Kartell™ PMMA (Plexiglas) Two-Tier Test Tube Racks feature smaller holes in the lower tier of the rack to offer stability while safely holding test tubes.

2435.00 NOK - 2740.00 NOK

Specifications

Product Type Test Tube Rack (2-tier)
Color Clear
Material PMMA
Positions 12
For Use With (Application) Smaller holes in the lower tier of the rack hold the test tubes safely
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12599716
View Documents
Kartell™
0088900
12 x 28 mm holes
2470.00 NOK
Pack of 5
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
12519726
View Documents
Kartell™
0089100
12 x 40 mm holes
2740.00 NOK
Pack of 5
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
12509726
View Documents
Kartell™
0089000
12 x 35 mm holes
2435.00 NOK
Pack of 5
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
Description

Description

  • Sturdy
  • Made of glass clear PMMA (Plexiglas)
  • Smaller holes in the lower tier of the rack hold the test tubes safely.
Specifications

Specifications

Test Tube Rack (2-tier)
PMMA
Smaller holes in the lower tier of the rack hold the test tubes safely
Clear
12
81 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ PMMA (Plexiglas) Two-Tier Test Tube Racks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.