Learn More
Brand™ PMMA Macro Cuvettes
Standard macro and micro cuvettes with a clear, clean optical path that indicates the optical path orientation.
Brand: Brand™ 759105
148.37 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- Macro and semi micro cuvettes
- Sorted by mould cavity number
- PMMA or PS
- Minimal extinction value variation
- Optically perfect transmission range
- Recessed window to protect against scratches
- Arrow head marking shows the direction of transmission
- Clear, clean optical path with indication of optical path orientation
- Manufactured under controlled room conditions and packaged fully automatically, without human contact
Specifications
Macro Cuvette | |
2.5 to 4.5 mL | |
PMMA | |
100Pack |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000014250 | Certificate of Analysis | 06/06/2025 | 759105 |
0000013827 | Certificate of Analysis | 05/05/2025 | 759105 |
0000013490 | Certificate of Analysis | 04/04/2025 | 759105 |
0000013395 | Certificate of Analysis | 10/03/2025 | 759105 |
0000013260 | Certificate of Analysis | 07/02/2025 | 759105 |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30