Learn More
Memmert™ Plug-In Tube Extension for SN75plus, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ SN75plusB29718
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
60.3 mm | |
SN75plus Sterilizer |
Product Content Correction
Product Title
Memmert™ Plug-In Tube Extension for SN75plus, Straight > For Use With: SN75plus