Learn More
Memmert™ Plug-In Tube Extension for SN55, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ SN55B29718
Product Code. 17538665
  • 554.00 NOK / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
60.3 mm
SN55 Sterilizer