Learn More
Memmert™ Plug-In Tube Extension for SF30, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ SF30B29718
Product Code. 17528215
  • 554.00 NOK / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
60.3 mm
SF30 Sterilizer
Product Title
Memmert™ Plug-In Tube Extension for SF30, Straight > For Use With: SF30