Learn More
Hecht Karl™ Plexiglas Test Tube Racks
Brand:  Hecht Karl™ 42807010
 View more versions of this product
Product Code. 10503652
  • 236.00 NOK / Each
Only null left
Estimated Shipment: 15-07-2025
Specifications
Test Tube Rack
Plexiglas
1 Pack
13 mm
Tubes (13 mm)
12
76 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hecht Karl™ Plexiglas Test Tube Racks > Height: 76mm