missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLDN Mouse anti-Human, Mouse, Rat, Clone: 4F4C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66558-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene may play a role in intracellular vesicle trafficking. It interacts with Syntaxin 13 which mediates intracellular membrane fusion. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
PLDN | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8A6, Q9R0C0, Q9UL45 | |
Bloc1s6 | |
PLDN Fusion Protein Ag18095 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG1 |
Western Blot | |
4F4C11 | |
Unconjugated | |
BLOC1S6 | |
PA, PALLID, Pallid protein homolog, Pallidin, pallidin homolog (mouse), PLDN | |
Mouse | |
Protein G | |
RUO | |
18457, 26258, 317630 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction