Learn More
Platinum Evaporating Basin with Flat Bottom
Platinum labware such as the Platinum Evaporating Basin with Flat Bottom features excellent properties that include high temperature strength and a high melting point, as well as ductility and corrosion and oxidation resistance..
Brand:  N/A 8411 3035
 View more versions of this product
Product Code. 12787366
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Flat bottom
  • Platinum
  • Heat resistant
Specifications
Specifications
Flat
Platinum
25 mm
35 mL
Evaporating Basin
50 mm