Learn More
Platinum, 5% on alumina paste, 5R94 Product Code.: 11374437
Platinum, 5% on alumina paste, 5R94
Product Code. 11374437
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11374437
Brand: Thermo Scientific Alfa Aesar 038318.06
Request Bulk or Custom Format
CAS: 7440-06-4 | Pt | 195.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
Specifications
Color Black
Quantity 5 g
Chemical Name or Material Platinum, 5% on alumina paste, 5R94
Product Title
Platinum, 5% on alumina paste, 5R94 >