missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Plate Count Agar, PCA, 100ml Flasks

Used as a medium for aerobic plate counts by the surface inoculation method (standard Plate Count Agar).

Brand:  Scharlab 064-BA1005

Product Code. 11439332

  • 544.00 NOK / Pack of 10
Estimated Shipment: 16-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Prepared according to ISO 4833:2003, 8552:2004, 17410:2001 and IFU No. 6 standards. (BAM / compf / idf / iFU / Iso / USP)

Specifications

Specifications

100mL Flasks
10 x 100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scharlab Plate Count Agar, PCA, 100ml Flasks >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.