Learn More
Kartell™ Plastilab™ Tapered Straight Connectors
For straight connection of tubing with different diameters.
Brand: Kartell™ 7051200
63.94 NOK valid until 2024-11-15
Use promo code "21643" to get your promotional price.
This item is not returnable.
View return policy
Description
For straight connection of tubings with different diameters.
- Constructed from polypropylene
- Autoclavable
Specifications
Straight Tapered Tubing Connector | |
Autoclavable | |
For straight connection of different diameter tubing | |
4.5mm bore |
8-10 mm | |
Polypropylene | |
10/Pk. | |
Straight |