Learn More
Hanna Instruments™ Plastic Pipette Tips
Used with the HI731341 Graduated Pipette. Hanna Instruments™ Plastic Pipette Tips are a 25-piece set of 1000μL disposable tips designed to precisely measure and transfer fixed volumes of liquids. Liquid is aspirated into disposable tips, eliminating cross contamination of samples.
Brand:  Hanna Instruments™ HI731351
Product Code. 11504527
  • 435.00 NOK / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Together, the HI731341 Graduated Pipette and HI731351 Plastic Pipette Tips are able to accurately transfer 1000μL volumes of liquid that need to be precisely dosed. By simply pressing the pushbutton to the first stop, immersing the disposable tip into the sample, and slowly releasing the pushbutton, an accurate volume of liquid is taken up. To dispense the liquid, press the pushbutton to the second stop. The built-in tip ejector provides a hassle-free way for users to discard their plastic tips.
  • 25 piece set
  • 1000μL volume
  • Durable plastic
  • Disposable
Specifications
Specifications
1000 μL
≤0.5%
25 pc.
HI731341 Graduated Pipette
Plastic
Natural
±1.2%
Single-use
Yes