Learn More
Plasmid DNA isolation solution II, Thermo Scientific™
311.00 NOK - 530.00 NOK
Specifica
Chemical Name or Material | Plasmid DNA isolation solution II |
---|---|
Color | Colorless |
Physical Form | Liquid |
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Plasmid DNA isolation solution II | |
Colorless | |
Liquid |
Sicurezza e movimentazione
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.