Learn More
Tocris Bioscience™ PKI 166 hydrochloride
Potent EGFR-kinase inhibitor
Brand: Tocris Bioscience™ 4941/10
1548.98 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
PKI 166 hydrochloride is a potent EGFR-kinase inhibitor (IC50 = 0.7 nM). Displays >3000-fold selectivity against a panel of serine/threonine kinases. Reduces metastasis and angiogenesis in a mouse model of pancreatic cancer. Antihypertensive and orally bioavailable.Specifications
>98% | |
10 mg | |
366.84 |
2230253-82-2 | |
PKI 166 hydrochloride | |
PKI 166 hydrochloride |
For Research Use Only