Learn More
Tocris Bioscience™ PKI 166 hydrochloride
Potent EGFR-kinase inhibitor
Brand:  Tocris Bioscience™ 4941/10
1548.98 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18748688
  • 1750.00 NOK / 10mg
Explore more special offers
Description
Description
PKI 166 hydrochloride is a potent EGFR-kinase inhibitor (IC50 = 0.7 nM). Displays >3000-fold selectivity against a panel of serine/threonine kinases. Reduces metastasis and angiogenesis in a mouse model of pancreatic cancer. Antihypertensive and orally bioavailable.
Specifications
Specifications
>98%
10 mg
366.84
2230253-82-2
PKI 166 hydrochloride
PKI 166 hydrochloride