missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PJA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17687-1-AP-150UL
This item is not returnable.
View return policy
Description
Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome. May be involved in protein sorting.Specifications
PJA1 | |
Polyclonal | |
Unconjugated | |
PJA1 | |
PJA1, praja ring finger 1, Praja1, RING finger protein 70, RNF70 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18744, 64219, 683077 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O55176, Q8NG27 | |
Pja1 | |
PJA1 Fusion Protein Ag11820 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction