missing translation for 'onlineSavingsMsg'
Learn More

PISD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen PISD
Concentration 0.21 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886014
View Documents Promotion Details
Proteintech
16401-1-AP-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16876014
View Documents Promotion Details
Proteintech
16401-1-AP-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to PISD. The sequence corresponds to the beta isoform.

Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
Specifications

Specifications

PISD
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Human, Rat
D3ZAW2, Q8BSF4, Q9UG56
23761, 320951, 681361
PISD Fusion Protein Ag9499
Primary
-20°C
PISD
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DJ858B16, PISD, PSDC, PSSC
Pisd
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications