missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroctone olamine, MedChemExpress

Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Piroctone olamine is a pyridine derivate. It is known to have a fungicidal effect.
Specifications
Specifications
Chemical Name or Material | Piroctone olamine |
CAS | 68890-66-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C16H30N2O3 |
Quantity | 50 mg |
Synonym | Piroctone ethanolamine |
Solubility Information | H2O : 50 mg/mL (167.55 mM; ultrasonic and adjust pH to 9 with NaOH) ∣DMSO : 11.11 mg/mL (37.23 mM; Need ultrasonic) |
SMILES | O=C1C=C(C)C=C(CC(C)CC(C)(C)C)N1O.NCCO |
Molecular Weight (g/mol) | 298.42 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Piroctone olamine, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction