Learn More
Eppendorf™ Pipette Stand 2, for Multipette™ M4
Saves bench space and conveniently stores your pipette for easy access.
Brand: Eppendorf™ 3116000058
1464.96 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Carries Eppendorf Multipette™ M4
- Robust design that securely holds and protects the instrument
- Large rubber feet protect stand from liquids spilled on workbench
Specifications
Pipette Holder | |
Multipipette M4 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%