Learn More
Socorex™ Pipette Nozzle Protection Filter
Introduce the Socorex™ Pipette Nozzle Protection Filter to protect the pipette from liquid fill or contamination. Made of polypropylene fiber, the filter offers maximal protection and safety from contamination.
525.00 NOK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Polypropylene Fiber |
Sterility | Non-sterile |
Type | Fiber Filter |
For Use With (Equipment) | Calibra™ digital 832, Acura™ manual 835, Acura™ electro 935/936 macroipettes and old Acura™ analogue 831 model |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17840051
|
Socorex™
322.05 |
Calibra™ digital 832, Acura™ manual 835, Acura™ electro 935/936 macroipettes and old Acura™ analogue 831 model |
525.00 NOK
Pack of 50 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
17850051
|
Socorex™
322.10 |
Calibra™ digital 832, Acura™ manual 835, Acura™ electro 935/936 macroipettes and old Acura™ analogue 831 model |
525.00 NOK
Pack of 50 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
Description
- Polypropylene fiber
- For 5mL and 10mL Acura™ manual and Acura™ electro
- Non sterile
- Non autoclavable.
Specifications
Not Autoclavable | |
Non-sterile | |
Calibra™ digital 832, Acura™ manual 835, Acura™ electro 935/936 macroipettes and old Acura™ analogue 831 model |
Polypropylene Fiber | |
Fiber Filter |