Learn More
Miele Professional™ Mobile Unit E 404/1
Mobile unit E 404/1 for the optimum loading of up to 38 pipettes
Brand: Miele Professional™ 6727910
This item is not returnable.
View return policy
Description
- Suitable for 38 pipettes in 3 rows
- 1st Row: 10 pipettes x 100 ml (up to 550 mm), bar distance 20 mm
- 2nd Row: 14 pipettes x 25 ml, bar distance 26 mm
- 3rd Row: 14 pipettes x 10 ml, bar distance 26 mm
- Without bracket for magnetic strip for mobile unit recognition
Specifications
Holds 38 pipettes | |
1Each | |
E 404/1 Mobile injector | |
Pipettes |