Learn More
Thermo Scientific™ Mitochondria Isolation Kit for Cultured Cells Product Code.: 10579663
Thermo Scientific™ Mitochondria Isolation Kit for Cultured Cells
Product Code. 10579663 Shop All Thermo Scientific Products
115 mL, 1 set
Click to view available options
Quantity:
115 mL
Unit Size:
1 set
Product Code. 10579663
Brand: Thermo Scientific™ 89874
Seek Out Great Savings With the Lab of Curiosity
Use promo code 24110 at checkout
View Available Promotions
Edge
Isolate intact mitochondria from cultured mammalian cell samples in about 40 minutes with this versatile, benchtop microcentrifuge-based fractionation kit.
Thermo Scientific™ Mitochondria Isolation Kit for Cultured Cells provides a versatile, microcentrifuge-tube method for fractionating intact mitochondria from cultured mammalian cell samples in about 40 minutes.
Isolation of mitochondria is typically a laborious process requiring single-sample processing with Dounce homogenization. The Mitochondria Isolation Kit uses a non-mechanical, reagent-based method that allows multiple (six) cultured cell samples to be processed concurrently in about 40 minutes. Cultured mammalian cell pellets are gently lysed using a proprietary formulation that results in maximum yield of mitochondria with minimal damage to integrity. The product instructions describe guidelines for optimizing purity vs. yield parameters. The kit also offers an optimized Dounce homogenization procedure, which results in two-fold greater mitochondria recovery compared to the reagent-based method. Both methods use differential centrifugation to separate the mitochondrial and cytosolic fractions with a benchtop microcentrifuge and are completed in approximately 40 minutes (post-cell harvest). Once isolated, the mitochondria can be used in downstream applications such as apoptosis, signal transduction and metabolic studies, as well as to facilitate mitochondrial proteomics efforts.
Highlights:
  • Fast – isolate intact mitochondria from 2 Ă— 107 cells in approximately 40 minutes (post-cell harvest)
  • Flexible – process several samples simultaneously using the reagent-based method, or obtain highest possible yield with the Dounce homogenization method
  • Optimizable – guidelines provided for optimizing purity vs. yield with the reagent-based method
  • Benchtop-easy – isolation performed in a microcentrifuge tube
  • Compatible – isolated mitochondria may be lysed with detergent or processed for almost any downstream application, including 2D electrophoresis
Includes:
Three buffer and detergent solutions totaling approx. 115mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cell Isolation Kit
Content And Storage Upon receipt store kit at 4°C. Product is shipped at ambient temperature.
Form Liquid
Reagent Type Organelle Isolation, Subcellular Fractionation
Sufficient For 50 Isolations
Quantity 115 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberAA403802 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code89874
Lot NumberZL403733A Certificate TypeCertificate of Analysis Date31/01/2025 Product Code89874
Lot NumberZL403733 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code89874
Lot NumberAA403802A Certificate TypeCertificate of Analysis Date31/01/2025 Product Code89874
Lot NumberZJ402170 Certificate TypeCertificate of Analysis Date16/12/2024 Product Code89874
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ Mitochondria Isolation Kit for Cultured Cells >