Learn More
Thermo Scientific™ MA(PEG)24 Methyl-PEG-Amine Compound Product Code.: 10617524
Thermo Scientific™ MA(PEG)24 Methyl-PEG-Amine Compound
Product Code. 10617524 Shop All Thermo Scientific Products
1 g, 1g
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 10617524
Brand: Thermo Scientific™ 26117
MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.
Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.
MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.
Highlights:
  • Pure compounds of specific size ensure complete control of PEGylation
  • Straight-chain compounds with simple terminal primary amine
  • Adaptable to a variety of modification procedures
Recommended for:
  • PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Reactivity Ketone
Description Pierce MA(PEG)24
Content And Storage Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
Label Type Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety Amine, Primary Amine
Spacer 24 ethylene glycol units
Form Solid
Quantity 1 g
Solubility Water
Product Type Amine Compound
Manuals
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberYH383107 Certificate TypeCertificate of Analysis Date01/09/2023 Product Code26117
Lot NumberYH381949 Certificate TypeCertificate of Analysis Date10/08/2023 Product Code26117
Lot NumberYD377325 Certificate TypeCertificate of Analysis Date22/04/2023 Product Code26117
Lot NumberYC375761 Certificate TypeCertificate of Analysis Date03/04/2023 Product Code26117
Lot NumberXJ367819 Certificate TypeCertificate of Analysis Date11/11/2022 Product Code26117
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ MA(PEG)24 Methyl-PEG-Amine Compound >