Learn More
Thermo Scientific™ Pierce™ Glutathione Superflow Agarose
Shop All Thermo Scientific ProductsDescription
Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Additional Information and Online Ordering
Highlights:
High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate
High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour
Compatible – maintains function after exposure to a wide variety of chemicals and pH values
Cost effective – resin is stable through multiple cycles of cleaning and reuse
Recommended for:
Medium- to large-scale FPLC purification of GST-tagged proteins
Specifications
Specifications
Description | Pierce Glutathione Superflow Agarose |
Formulation | Glutathione onhighly crosslinked 6% beaded agarose (superflow 6), slurried in water with sodium azide |
Quantity | 1 L |
Sufficient For | Binding up to 30mg GST/mL resin (static capacity) or up to 10mg GST/mL resin (dynamic capacity) |
Documents
Product Information
Manuals