Learn More
Thermo Scientific™ Pierce™ 1-Step Transfer Buffer Product Code.: 15175204
Thermo Scientific™ Pierce™ 1-Step Transfer Buffer
Product Code. p-6019434 Shop All Thermo Scientific Products
1 L, 1 L
Click to view available options
Quantity:
200 mL
1 L
5 x 1 L
Unit Size:
1 L
200mL
5 L
Product Code. 15175204
Brand: Thermo Scientific™ 84731
Perform rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
The Thermo Scientific™ Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
Thermo Scientific Pierce 1-Step Transfer Buffer is designed for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Power Blotter.
Features of 1-Step Transfer Buffer:
Fast—high ionic strength formulation provides 5- to 10-minute protein transfer when used with compatible fast semi-dry blotting systems
Ready to use—supplied as a 1X solution that is stable and ready to use at room temperature
Optimized—designed for seamless and reliable performance with the Power Blotter
Compatible—effective with other semi-dry transfer devices equipped with a suitable high-current power supply, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217)
Economical—excellent performance without the special or costly consumables that are required by other fast semi-dry transfer devices
When used with the Power Blotter (Cat. No. 62288), this buffer provides effective gel-to-membrane transfer of proteins in 5 to 10 minutes with efficiency that is equal to, or better than, conventional Western blotting techniques. Pierce 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices, including the original Pierce Fast Semi-Dry Blotter (Part No. 88217), when they are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.
Requires:
Power Blotter (Part No. 62288) or other semi-dry transfer device that is equipped or paired with a capable high-current power supply.
Fast blotting methods require a high-current power supply, such as the Power Blotter, and an optimized high ionic strength transfer buffer, such as Pierce 1-Step Transfer Buffer. By increasing the current, excellent transfer efficiency can be achieved in much shorter time compared to conventional methods. Amperage is held at a constant rate based on the surface area of the transfer stack(s) (∼22-23 mA/cm2) and voltage is limited to 25 V.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Transfer Buffer
Buffer Transfer Buffers
Concentration 1 X
Content And Storage Upon receipt store at room temperature. Product is shipped at ambient temperature.
Gel Type SDS-PAGE Gel (any)
Gel Compatibility SDS-PAGE Gel
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce™
Quantity 1 L
  • Thermo Scientific - Pierce G2 Fast Blotter
    The Thermo Scientific Pierce G2 Fast Blotter is designed specifically for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 10 minutes.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific Pierce G2 Fast Blotter
    • Thermo Scientific - Pierce G2 Fast Blotter
      The Thermo Scientific Pierce G2 Fast Blotter is designed specifically for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 10 minutes.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific Pierce G2 Fast Blotter
      Certificates
      A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
      5 results found
      Lot Number Certificate Type Date Product Code
      Lot NumberZH392070 Certificate TypeCertificate of Analysis Date10/09/2024 Product Code84731
      Lot NumberYJ380226 Certificate TypeCertificate of Analysis Date07/12/2023 Product Code84731
      Lot NumberYA364659 Certificate TypeCertificate of Analysis Date10/02/2023 Product Code84731
      Lot NumberXF346448 Certificate TypeCertificate of Analysis Date30/08/2022 Product Code84731
      Lot NumberWI331851 Certificate TypeCertificate of Analysis Date09/11/2021 Product Code84731
      For Research Use Only. Not for use in diagnostic procedures.
      Product Content Correction
      Product Title
      Thermo Scientific™ Pierce™ 1-Step Transfer Buffer >