Learn More
Piceatannol, MedChemExpress
MedChemExpress Piceatannol is a well-known Syk inhibitor and reduces the expression of iNOS induced by TNF.
Brand: Medchem Express HY-13518
This item is not returnable.
View return policy
Description
Piceatannol is an effective agent for research of acute lung injury (ALI). Piceatannol is a naturally occurring polyphenolic stilbene found in various fruits and vegetables and exhibits anticancer and anti-inflammatory properties. Piceatannol induces apoptosis in DLBCL cell lines. Piceatannol induces autophagy and apoptosis in MOLT-4 human leukemia cells.Specifications
Piceatannol | |
Earth Yellow | |
C14H12O4 | |
Astringenin trans-Piceatannol | |
OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C=C1O | |
244.24 | |
Research | |
Cancer-Kinase/protease |
10083-24-6 | |
Research | |
25 mg | |
DMSO : ≥ 31 mg/mL (126.92 mM) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic) | |
244.24 | |
98.09% | |
Solid |