missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PI3 (Human) Recombinant Protein

Human PI3 full-length ORF ( AAH10952, 1 a.a. - 117 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005266-P01.10ug

 View more versions of this product

Product Code. 16139061

  • 4220.00 NOK / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Sequence: MRASSFLIVVVFLIAGTLVLEAAVTGVPVKGQDTVKGRVPFNGQDPVKGQVSVKGQDKVKAQEPVKGPVSTKPGSCPIILIRCAMLNPPNRCLKDTDCPGIKKCCEGSCGMACFVPQ
Specifications

Specifications

AAH10952
peptidase inhibitor 3, skin-derived
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PI3
GST
5266
Wheat germ expression system
10 μg
ESI, MGC13613, SKALP, WAP3, WFDC14, cementoin
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ PI3 (Human) Recombinant Protein > Quantity: 10 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.