missing translation for 'onlineSavingsMsg'
Learn More
Phorbol 12-myristate 13-acetate, ≥99%, MP Biomedicals™ Product Code.: 11422131

Phorbol 12-myristate 13-acetate, ≥99%, MP Biomedicals™

Product Code. 11422131 Shop All MP Biomedicals Products
1 mg
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy

Product Code. 11422131

Brand: MP Biomedicals 0219480401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 16561-29-8
Color White to Yellow
Molecular Formula C36H56O8
MDL Number MFCD00036736
Quantity 1 mg
Synonym phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate
InChI Key PHEDXBVPIONUQT-RGYGYFBISA-N
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
IUPAC Name (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Weight (g/mol) 616.84
PubChem CID 27924
ChEBI CHEBI:37537
Formula Weight 616.84
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Phorbol 12-myristate 13-acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phorbol 12-myristate 13-acetate, ≥99%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.