missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phorbol 12-Myristate 13-acetate, ∽99%, Molecular biology reagent grade, MP Biomedicals™
Click to view available options
Quantity:
10 mg
Specifications
Specifications
CAS | 16561-29-8 |
Assay Percent Range | Approx. 99% |
Molecular Formula | C36H56O8 |
MDL Number | MFCD00036736 |
Quantity | 10 mg |
Synonym | phorbol 12-myristate 13-acetate, phorbol ester, 12-o-tetradecanoylphorbol-13-acetate, 12-o-tetradecanoylphorbol 13-acetate, factor a1, tetradecanoylphorbol acetate, 12-tetradecanoylphorbol 13-acetate, factor a1 croton oil, phorbol 13-acetate 12-myristate, phorbol-12-myristate-13-acetate |
InChI Key | PHEDXBVPIONUQT-RGYGYFBISA-N |
SMILES | CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O |
IUPAC Name | (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate |
Molecular Weight (g/mol) | 616.84 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'