missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Phenyl-Sil Deactivated Capillary Column
Brand: Macherey-Nagel™ 723348.25
This item is not returnable.
View return policy
Specifications
0.5 mm | |
0.32 mm | |
320°C | |
1 Each |
Deactivated Capillary Column | |
Preparation of Capillary Columns Analysis | |
25 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Phenyl-Sil Deactivated Capillary Column > I.D.: 0.32mm; Length: 25m
Spot an opportunity for improvement?Share a Content Correction