missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Isocyanate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 103-71-9 |
Molecular Formula | C7H5NO |
Quantity | 100 g |
Synonym | phenyl isocyanate, phenylcarbimide, carbanil, phenyl carbonimide, benzene, isocyanato, phenylisocyanate, isocyanic acid, phenyl ester, mondur p, phnco, fenylisokyanat |
InChI Key | DGTNSSLYPYDJGL-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)N=C=O |
IUPAC Name | isocyanatobenzene |
Molecular Weight (g/mol) | 119.123 |
PubChem CID | 7672 |
ChEBI | CHEBI:53806 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'