Learn More
Phenolic Solution, Chem Lab™
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Quantity | 100 mL |
Color | Colorless |
Odor | Specific |
pH | >10 |
Solubility | Soluble in water |
Physical Form | Liquid |