missing translation for 'onlineSavingsMsg'
Learn More
pHenol Disulfonic Acid Solution 25%(w/v) In Sulfuric Acid, Pure, Fisher Chemical Product Code.: 10766291

pHenol Disulfonic Acid Solution 25%(w/v) In Sulfuric Acid, Pure, Fisher Chemical

Product Code. 10766291
500 mL, Plastic Coated Glass bottle
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10766291

Brand: Fisher Chemical J/5900/PB08

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Color White
Packaging Plastic Coated Glass bottle
MDL Number 152742
Quantity 500 mL
Physical Form Liquid
compliance-icons
Product Identifier
  • Phenol disulfonic acid solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pHenol Disulfonic Acid Solution 25%(w/v) In Sulfuric Acid, Pure, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.