Learn More
PHACS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11491-1-AP-20UL
 View more versions of this product
Product Code. 16898603
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
Does not catalyze the synthesis of 1-aminocyclopropane- 1-carboxylate but is capable of catalyzing the deamination of L- vinylglycine.
Specifications
Specifications
PHACS
Polyclonal
Unconjugated
ACCS
ACC synthase like protein 1, ACCS, ACS, PHACS
Rabbit
Antigen Affinity Chromatography
RUO
311218, 329470, 84680
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2AIG8, Q96QU6
Accs
PHACS Fusion Protein Ag2056
20 μL
Primary
Mouse, Rat, Human