Description
- 6 pHast Clean™ Electrode
- The pHast Clean electrode makes it possible to renew the junction by forcing the passage of the electrolyte by pressure on the head of the electrode
- This system increases the life of the electrode, reduces cleaning operations and stabilizes the measured items
- Ideal for loaded or viscous solutions
- Electrolyte KCl 4M + AgCl
Specifications
BNC | |
pH Electrode | |
Glass | |
0°C to 80°C |
12 mm | |
120 mm | |
0 to 14 |