Learn More
pH Buffer Solution NBS DIN 19226 pH 6.8, Mettler Toledo™
GREENER_CHOICE
Brand:  Mettler Toledo 51350054
300.47 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Quantity 250 mL
Product Code. 10474071
  • 349.00 NOK / 250mL
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Edge
Specifications
6.8
250 mL
1Each
Yes
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
Product Content Correction
Product Title
pH Buffer Solution NBS DIN 19226 pH 6.8, Mettler Toledo™ > Quantity: 250mL